Dräger™ CMS Chips: Hydrogen Sulfide
Suitable for 10 measurements of one gas.
Manufacturer:  Dräger™ 6406220
 View more versions of this product
Catalog No. 29-801-246
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100hpa
100 to 2500
Hydrogen Sulphide CMS™ chip
0° to 40°C
1 to 40mg/L
40 to 500 sec.
32° to 104°F