Dräger™ CMS Chips: Carbon Dioxide
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406210
 View more versions of this product
Catalog No. 29-801-245
Description
Description
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
Safety
1 to 30mg/L
12 to 120 sec.
32° to 104°F
700 to 1100hpa
1 to 20%
Carbon Dioxide CMS™ chip
0° to 40°C
Documents