Dräger™ CMS Chips: Carbon Dioxide
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406190
 View more versions of this product
Catalog No. 29-801-244
Description
Description
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100hpa
200 to 3000
Carbon Dioxide CMS™ chip
0° to 40°C
1 to 30mg/L
60 to 260 sec.
32° to 104°F