Dräger™ CMS Chips: Benzene
Suitable for 10 measurements of one gas
Manufacturer:  Dräger™ 6406280
 View more versions of this product
Catalog No. 29-801-243
Description
Description
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
Safety
1 to 30mg/L
40 to 275 sec.
32° to 104°F
700 to 1100hpa
10 to 250
Benzene CMS™ chip
0° to 40°C
Documents