Dräger™ accuro™ Bellows Pump: Hardside Kit
Ergonomic design allows easy measurement of more than 350 gases—including formaldehyde
Manufacturer:  Dräger™ 4056443
Catalog No. 17-988-409
Includes: Contains the accuro pump, deluxe tube opener, a pack of rubber caps for tubes, a screwdriver, an Allen wrench, and spare parts for the pump, hard plastic carrying case.
Description
Description
Provides a means of direct, onsite measuring of more than 350 gases, vapors, and aerosols. Internal scissors mechanism provides stability so that a uniform, even pump stroke of 100cm3 can be completed.
  • Convenient, built-in stroke counter records strokes performed
  • Recessed counter reset button prevents accidental resetting; a white dot in clear optical window of pump rises to indicate completion of each stroke cycle, preventing incomplete measurements
  • Ergonomic design fits comfortably in one hand
  • Soft rubber grips on pump body make it easier to hold and squeeze
  • Built-in ceramic tube opener adds safety and convenience Corrosion-resistant construction provides years of reliable service
  • Components are easily replaceable and snap together for quick reassembly
  • Use with accessory extension hoses for remote sampling
  • Designed and manufactured using ISO 90001:2008-certified processes
Order Info
Pump dimensions: 6.75L x 1.75W x 3.75 in.H (17.1 x 4.5 x 8.5cm). Net weight: 0.5 lb. (225g). Stroke volume: 100cm3 ±5%. Softside Kit, pump only and hard case available separately (Cat. Nos. 17-988-408, 17-986-70A and 19-046-749).
Specifications
Specifications
Accuro™ pump, deluxe tube opener, Rubber caps for tubes, screwdriver, Allen wrench, spare parts for pump, hard Plastic carrying case
Accuro™ hardside kit