Please login to your online account to display your discounted pricing

Dr. FuelCell™ Science Kits

Hands-on experience using individual investigations or small-group activities to demonstrate a solar hydrogen system.

$197.10 - $1,034.10

Specifications

Grade 9 to 12
Includes Solar panel, PEM electrolyzer, PEM fuel cell, load measurement box, four-volume lesson book set, Clean Energy for the Future CD-ROM, illustrated set-up sheets, stopwatch, cables, plastic storage bin, and dismantleable fuel cell and methanol fuel cell ex
Storage Requirements 10° to 35°C
View More Specs
Includes:  Solar panel, PEM electrolyzer, PEM fuel cell, load measurement box, four-volume lesson book set, Clean Energy for the Future CD-ROM, illustrated set-up sheets, stopwatch, cables, plastic storage bin, and dismantleable fuel cell and methanol fuel cell extensions (complete kit only)
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S64254

 
fisher scientific
350EMD*
Basic Kit Each for $511.65

S65851

 
fisher science education
357E
Methanol Fuel Cell Extension Kit Each for $197.10

S97303

 
fisher science education
K0355E
Fuel Kit Each for $1,034.10
Description & Specifications

Specifications

Grade 9 to 12
Includes Solar panel, PEM electrolyzer, PEM fuel cell, load measurement box, four-volume lesson book set, Clean Energy for the Future CD-ROM, illustrated set-up sheets, stopwatch, cables, plastic storage bin, and dismantleable fuel cell and methanol fuel cell ex
Storage Requirements 10° to 35°C

  • Demonstrates the sustainable benefits of fuel cells and hydrogen technology to introductory and advanced high school courses
  • Electrolyzer uses only distilled water for safe and easy setup
  • Covers key curriculum topics with 25 experiments, 19 readings and background information
  • Load measurement box has a voltmeter and ammeter, two electrical loads (lamp and electric motor) and seven different adjustable resistances
  • Add more experiments and a deeper understanding of fuel cell technology with extension kits
  • Grades 9-12
Specifications: For detailed specifications of solar panel, PEM electrolyzer, load measurement box and fuel cells, see www.fisheredu.com.