Please login to your online account to display your discounted pricing

Dr. FuelCell™ Science Kits


Hands-on experience using individual investigations or small-group activities to demonstrate a solar hydrogen system.

Manufacturer: fisher science education  357E

 View more versions of this product

Catalog No. S65851

  • / Each
 

Description & Specifications

Specifications

Grade 9 to 12
Includes Solar panel, PEM electrolyzer, PEM fuel cell, load measurement box, four-volume lesson book set, Clean Energy for the Future CD-ROM, illustrated set-up sheets, stopwatch, cables, plastic storage bin, and dismantleable fuel cell and methanol fuel cell ex
Storage Requirements 10° to 35°C
Product Type Methanol Fuel Cell Extension Kit