Please login to your online account to display your discounted pricing

Dr. FuelCell™ Model Cars


Teach concepts such as electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps.

Manufacturer: fisher science education  352E

 View more versions of this product

Catalog No. S65853

  • / Each
 

Description & Specifications

Specifications

Grade 8 to 12
Storage Requirements 10° to 35°C
For Use With (Application) Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps
Product Type Demo Kit
Includes Car chassis with electric motor,fuel cell, solar panel, car cassis, storage case, cables, and lesson book