Please login to your online account to display your discounted pricing

Dr. FuelCell™ Model Cars


Teach concepts such as electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps.

Manufacturer: fisher scientific  K921E

 View more versions of this product

Catalog No. S97302

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Product Type Classroom Package of 10 Demos
For Use With (Application) Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps
Includes 10 car chassis with electric motor, 10 fuel cells, 10 solar panels, 10 car chassis, 10 storage cases, cables, 2 lesson books, 4 load measurement box
Storage Requirements 10° to 35°C
Grade 8 to 12

  • Water and sunlight power a simple and effective demonstration of solar hydrogen technology
  • Reversible PEM fuel cell works as both an electrolyzer and a fuel cell
  • As an electrolyzer, it uses power from a solar module to split water into hydrogen and oxygen
  • As a fuel cell, it uses stored hydrogen and oxygen to power the electric motor and drive the car
  • Load measurement box has a voltmeter, ammeter, two electrical loads (lamp and electric motor) and seven different adjustable resistances for characteristic curve analysis
  • Course book is written by an award-winning teacher; contains eight experiments designed with teacher and student in mind
  • Grades 8-12
Specifications: For specifications, please visit www.fisheredu.com.