Please login to your online account to display your discounted pricing

Dr. FuelCell™ Model Cars


Teach concepts such as electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps.

Manufacturer: fisher scientific  K920E

 View more versions of this product

Catalog No. S97301

  • / Pack of 5 Each
 

Description & Specifications

Specifications

For Use With (Application) Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps
Grade 8 to 12
Includes 5 car chassis with electric motor, 5 fuel cell, 5 solar panel, 5 car chassis, 5 storage cases, cables, 2 lesson book, 2 load measurement box
Storage Requirements 10° to 35°C
Product Type Classroom Package of 5 Demos