Dr. FuelCell™ Model Cars
Teach concepts such as electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps.
Manufacturer:  Fisher Scientific 354E
 View more versions of this product
Catalog No. S66135
Description
Description
  • Water and sunlight power a simple and effective demonstration of solar hydrogen technology
  • Reversible PEM fuel cell works as both an electrolyzer and a fuel cell
  • As an electrolyzer, it uses power from a solar module to split water into hydrogen and oxygen
  • As a fuel cell, it uses stored hydrogen and oxygen to power the electric motor and drive the car
  • Load measurement box has a voltmeter, ammeter, two electrical loads (lamp and electric motor) and seven different adjustable resistances for characteristic curve analysis
  • Course book is written by an award-winning teacher; contains eight experiments designed with teacher and student in mind
  • Grades 8-12
Specifications: For specifications, please visit www.fisheredu.com.
Specifications
Specifications
Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps
Car chassis with electric motor, fuel cell, solar panel, car cassis, storage case, cables, and lesson book, load measurement box
Complete Kit
8 to 12
10° to 35°C
Documents