Please login to your online account to display your discounted pricing

DPPI 1c hydrochloride, Tocris

Dipeptidyl peptidase IV (DPP-IV) inhibitor

$133.98 - $284.81

Chemical Identifiers

Chemical Name or Material DPPI 1c hydrochloride
Assay Percent Range >98%
Molecular Formula C14H20N4O2·HCl
Formula Weight 312.8
Synonym 1-[[[(1-Hydroxymethyl)cyclopentyl]amino]acetyl]-2, 5-cis-pyrrolidinedicarbonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

278310

 
r & d systems
278310
10mg Each for $284.81

27831

 
r & d systems
27831
1mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 312.8
Quantity 10mg
Chemical Name or Material DPPI 1c hydrochloride
Assay Percent Range >98%
Molecular Formula C14H20N4O2·HCl
CAS 866396-70-5
Synonym 1-[[[(1-Hydroxymethyl)cyclopentyl]amino]acetyl]-2, 5-cis-pyrrolidinedicarbonitrile hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature