Please login to your online account to display your discounted pricing

DPPE fumarate, Tocris


Inhibitor of histamine binding at the intracellular binding site

Quantity 10mg

Manufacturer: r & d systems  RDS074310

 View more versions of this product

Catalog No. 074310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 399.49
CAS 1185241-83-1
Assay Percent Range >98%
Synonym N, N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine fumarate
Chemical Name or Material DPPE fumarate
Molecular Formula C19H25NO·C4H4O4
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material DPPE fumarate
Assay Percent Range >98%
Molecular Formula C19H25NO·C4H4O4
Formula Weight 399.49
Synonym N, N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine fumarate
Accessories

Desiccate at -20°C