Please login to your online account to display your discounted pricing

DPPE fumarate, Tocris

Inhibitor of histamine binding at the intracellular binding site

$159.50 - $669.90

Chemical Identifiers

Chemical Name or Material DPPE fumarate
Assay Percent Range >98%
Molecular Formula C19H25NO·C4H4O4
Formula Weight 399.49
Synonym N, N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine fumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

074350

 
r & d systems
RDS074350
50mg Each for $669.90

074310

 
r & d systems
RDS074310
10mg Each for $159.50
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 399.49
Chemical Name or Material DPPE fumarate
Assay Percent Range >98%
Molecular Formula C19H25NO·C4H4O4
Synonym N, N-Diethyl-2-[4-(phenylmethyl)phenoxy]ethanamine fumarate
CAS 1185241-83-1
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C