missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DPD No. 1 Rapid Tablets, Lovibond™

$42.00 - $83.69

Chemical Identifiers

Physical Form Tablets
Chemical Name or Material DPD No. 1 Rapid Tablets, for DPD Method
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09-054-302 Tintometer Inc.
511311BT
100 Tablets Pack of 250 Each for $83.69
36-104-1275 Tintometer Inc.
511310BT
100 Tablets Each for $42.00
Specifications Description & Specifications

Chemical Identifiers

Tablets
DPD No. 1 Rapid Tablets, for DPD Method

Specifications

100 Tablets
Tablets
DPD No. 1 Rapid Tablets, for DPD Method