Please login to your online account to display your discounted pricing

Doxylamine succinate, 98%, Acros Organics


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC449540500

 View more versions of this product

Catalog No. AC449540500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 388.46
Quantity 50g
CAS 562-10-7
Packaging Glass bottle
Chemical Name or Material Doxylamine succinate
Molecular Formula C17H22N2O·C4H6O4
Assay Percent Range 98%

Specifications

Chemical Name or Material Doxylamine succinate
Assay Percent Range 98%
Molecular Formula C17H22N2O·C4H6O4
Formula Weight 388.46