Please login to your online account to display your discounted pricing

Alfa Aesar™ Doxorubicin hydrochloride, 10mg/mL in DMSO, sterile filtered

Chemical Identifiers

Chemical Name or Material Doxorubicin hydrochloride
CAS 25316-40-9
Name Note 10mg/mL in DMSO, sterile filtered
MDL Number MFCD00077757
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67209XF

  SDS
alfa aesar™
J67209XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Doxorubicin hydrochloride
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00077757
CAS 25316-40-9
Health Hazard 1 Danger
Physical Form Liquid
Storage Note 1 -30° to -10°C
Health Hazard 2 GHS H Statement
H350
May cause cancer.
Name Note 10mg/mL in DMSO, sterile filtered
RTECS Number QI9295900
Quantity 1mL

Applications
Doxorubicin hydrochloride is a DNA intercalator and powerful cytotoxic agent. Doxorubicin inhibits topoisomerase II which results in an increased and stabilized cleavable enzyme-DNA linked complex during DNA replication and subsequently prevents the ligation of the nucleotide strand after double-strand breakage.

Solubility
Soluble in water (50 mM), methanol, isotonic sodium chloride, and DMSO (>10 mg/ml). Insoluble in chloroform, and ether.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350
May cause cancer.

-30° to -10°C