Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex Marathon C, H+ form

$81.12 - $309.39

Chemical Identifiers

Chemical Name or Material Dowex Marathon C
Name Note H+ form
MDL Number MFCD00212649
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB257210E

 
alfa aesar™
B257210E
2500g Each for $309.39

AAB2572130

 
alfa aesar™
B2572130
250g Each for $81.12

AAB257210B

 
alfa aesar™
B257210B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dowex Marathon C
TSCA TSCA
Name Note H+ form
MDL Number MFCD00212649
Quantity 2500g

Applications
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.