Please login to your online account to display your discounted pricing

Dowex™ 5WX2-200, ion-exchange resin, ACROS Organics™

$112.18 - $112.18

Chemical Identifiers

Chemical Name or Material Dowex™ 50WX2
CAS 12612-37-2
MDL Number MFCD00132722
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203030025

  SDS
thermo fisher scientific
AC203030025
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203031000

  SDS
fisher scientific acros
203031000
Plastic Bottle 100g Each for $112.18

AC203035000

  SDS
fisher scientific acros
203035000
Plastic Bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 2.5kg
Physical Form Beads
Chemical Name or Material Dowex™ 50WX2
Particle Size Particle size analysis: on 35 mesh (cumulative): 15% max., through 140 mesh: 15% max.
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 0.6
Water Retention Capacity H form: 74 to 82%
MDL Number MFCD00132722
CAS 12612-37-2