missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 50X4-400, ion-exchange resin, ACROS Organics™

Manufacturer:  Acros Organics 203051000

 View more versions of this product

Catalog No. AC203051000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Dowex™ 50WX4
Particle size analysis: on 100 mesh (cumulative): <15%, through 325 mesh: <15%
MFCD00132725
H form: 64 to 72%
Beads
11113-61-4
Plastic Bottle
wet volume capacity, H form (meq/mL):> 1.1
100g

Chemical Identifiers

11113-61-4
MFCD00132725
SDS Documents