Please login to your online account to display your discounted pricing

Dowex™ 50X2-400, ion-exchange resin, ACROS Organics™

$98.89 - $363.54

Chemical Identifiers

Chemical Name or Material Dowex™ 50WX2
CAS 12612-37-2
MDL Number MFCD00132722
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203040025

  SDS
thermo fisher scientific
AC203040025
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203041000

  SDS
thermo fisher scientific
203041000
Plastic Bottle 100g Each for $98.89

AC203045000

  SDS
thermo fisher scientific
203045000
Plastic Bottle 500g Each for $363.54
Description & Specifications

Specifications

Quantity 2.5kg
Physical Form Beads
Chemical Name or Material Dowex™ 50WX2
Particle Size Particle size analysis: on 70 mesh (cumulative): <15%, through 270 mesh: <15%
Water Volume Capacity wet volume capacity, H form (meq/mL)> 0.6
Water Retention Capacity H form: 74 to 82%
MDL Number MFCD00132722
CAS 12612-37-2