Please login to your online account to display your discounted pricing

Dowex™ 50X2-400, ion-exchange resin, ACROS Organics™


Quantity 500g
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  203045000

 View more versions of this product

Catalog No. AC203045000

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Physical Form Beads
Water Retention Capacity H form: 74 to 82%
Chemical Name or Material Dowex™ 50WX2
MDL Number MFCD00132722
Packaging Plastic Bottle
Particle Size Particle size analysis: on 70 mesh (cumulative): <15%, through 270 mesh: <15%
Water Volume Capacity wet volume capacity, H form (meq/mL)> 0.6
CAS 12612-37-2

Specifications

Chemical Name or Material Dowex™ 50WX2
CAS 12612-37-2
MDL Number MFCD00132722