Please login to your online account to display your discounted pricing

Dowex™ 50X2-100, ion-exchange resin, ACROS Organics™


Quantity 2.5kg

Manufacturer: thermo fisher scientific  AC203020025

 View more versions of this product

Catalog No. AC203020025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 2.5kg
Physical Form Beads
Fieser 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176
Chemical Name or Material Dowex™ 50WX2
Particle Size Particle size analysis: on 20 mesh (cumulative): <15%, through 70 mesh: <15%
Water Volume Capacity wet volume capacity, H form (meq/mL):> 0.6
Water Retention Capacity H form: 74 to 82%
MDL Number MFCD00132722
CAS 12612-37-2

Specifications

Chemical Name or Material Dowex™ 50WX2
CAS 12612-37-2
MDL Number MFCD00132722