missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 50X2-100, ion-exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203025000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beads | |
12612-37-2 | |
Plastic Bottle | |
MFCD00132722 | |
01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176 |
Dowex™ 50WX2 | |
Particle size analysis: on 20 mesh (cumulative): <15%, through 70 mesh: <15% | |
wet volume capacity, H form (meq/mL):> 0.6 | |
H form: 74 to 82% | |
500 g |
Chemical Identifiers
12612-37-2 | |
MFCD00132722 |
RUO – Research Use Only