missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 50WX8-400 ion-exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335350025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beads | |
11119-67-8 | |
Plastic bottle | |
MFCD00132726 | |
Beige to Brown | |
Solubility in water: insoluble. |
Dowex™ 50WX8 | |
15% max. (on 100 mesh, cumulative), 15% max. (through 325 mesh) | |
wet volume capacity, H form min. (meq/mL): 1.7 | |
H form: 50 to 58% | |
2.5 kg |
Chemical Identifiers
11119-67-8 | |
MFCD00132726 |
RUO – Research Use Only