Please login to your online account to display your discounted pricing

Dowex™ 50WX8-200 ion-exchange resin, ACROS Organics™


Quantity 100g
Packaging Plastic Bottle

Manufacturer: fisher scientific acros  335341000

 View more versions of this product

Catalog No. AC335341000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Beige to brownish
Physical Form Moist Beads or Spheres
Quantity 100g
Particle Size 15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
Packaging Plastic Bottle
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 1.7
CAS 11119-67-8
MDL Number MFCD00132726
Water Retention Capacity H form: 50 to 58%
Chemical Name or Material Dowex™ 50WX8

Specifications

Chemical Name or Material Dowex™ 50WX8
CAS 11119-67-8
MDL Number MFCD00132726