Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX8 200-400 (H)


Quantity 250g

Manufacturer: alfa aesar™  L1392230

 View more versions of this product

Catalog No. AAL1392230

  • / Each
 

Description & Specifications

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Dowex™ 50WX8 200-400 (H)
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 1 Warning
Quantity 250g

Specifications

Chemical Name or Material Dowex™ 50WX8 200-400 (H)
CAS 11119-67-8
MDL Number MFCD00132726

Applications
Dowex« 50WX8 200-400 (H) is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Accessories

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

SDS & Safety Documents