missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 50WX8-100 ion-exchange resin, ACROS Organics™

$116.93 - $396.69

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335335000 SDS View Documents Acros Organics
335335000
500g Plastic Bottle Each for $396.69
AC335331000 SDS View Documents Acros Organics
335331000
100g Plastic Bottle Each for $116.93
AC335330025 SDS View Documents Acros Organics
AC335330025
2.5kg Plastic bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

11119-67-8
MFCD00132726

Specifications

11119-67-8
Plastic Bottle
wet volume capacity, H form min. (meq/mL): 1.7
01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256
Solubility in water: insoluble.
Moist Spheres
15% max. (on 35 mesh, cumulative), 20% max. (through 80 mesh)
MFCD00132726
H form: 50 to 56%
Beige to Tan or Green-Tan
500g
Dowex™ 50WX8