Please login to your online account to display your discounted pricing

Dowex™ 50WX8-100 ion-exchange resin, ACROS Organics™


Quantity 2.5kg

Manufacturer: thermo fisher scientific  AC335330025

 View more versions of this product

Catalog No. AC335330025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Moist Spheres
Quantity 2.5kg
Color Beige to tan or green-tan
Fieser 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256
Particle Size 15% max. (on 35 mesh, cumulative), 20% max. (through 80 mesh)
Water Volume Capacity wet volume capacity, H form min. (meq/mL): 1.7
Water Retention Capacity H form: 50 to 56%
MDL Number MFCD00132726
CAS 11119-67-8
Chemical Name or Material Dowex™ 50WX8

Specifications

Chemical Name or Material Dowex™ 50WX8
CAS 11119-67-8
MDL Number MFCD00132726