Learn More
AmberChrom 50Wx8 100-200 (H), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1392130
Description
Dowex™ 50WX8 100-200 (H) is used as a separation medium for thin layer chromatography and low pressure liquid chromatography. It is also used in the separation process in chemicals and pharmaceuticals. Further, it is employed in deionizing supernatant fraction samples for paper chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDowex|r 50WX8 100-200 (H) is used as a separation medium for thin layer chromatography and low pressure liquid chromatography. It is also used in the separation process in chemicals and pharmaceuticals. Further, it is employed in deionizing supernatant fraction samples for paper chromatography.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Dowex™ 50WX8 100-200 (H) | |
MFCD00132726 | |
Insoluble in water. |
11119-67-8 | |
250 g |
Chemical Identifiers
11119-67-8 | |
MFCD00132726 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only