Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX8 100-200 (H)


Quantity 50g

Manufacturer: alfa aesar™  L1392118

 View more versions of this product

Catalog No. AAL1392118

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Dowex™ 50WX8 100-200 (H)
MDL Number MFCD00132726
CAS 11119-67-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Quantity 50g
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Specifications

Chemical Name or Material Dowex™ 50WX8 100-200 (H)
CAS 11119-67-8
MDL Number MFCD00132726

Applications
Dowex|r 50WX8 100-200 (H) is used as a separation medium for thin layer chromatography and low pressure liquid chromatography. It is also used in the separation process in chemicals and pharmaceuticals. Further, it is employed in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Accessories

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

SDS & Safety Documents