Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX4 200-400 (H)

$64.80 - $593.12

Chemical Identifiers

Chemical Name or Material Dowex™ 50WX4 200-400 (H)
CAS 11113-61-4
MDL Number MFCD00132725
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL139180B

 
alfa aesar™
L139180B
1000g Each for $593.12

AAL1391818

 
alfa aesar™
L1391818
50g Each for $64.80

AAL1391830

 
alfa aesar™
L1391830
250g Each for $199.23
Description & Specifications

Specifications

MDL Number MFCD00132725
CAS 11113-61-4
Chemical Name or Material Dowex™ 50WX4 200-400 (H)
Quantity 1000g

Applications
Dowex® 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex® 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.