Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX4 200-400 (H)


Quantity 1000g

Manufacturer: alfa aesar™  L139180B

 View more versions of this product

Catalog No. AAL139180B

  • / Each
 

Description & Specifications

Chemical Identifiers

MDL Number MFCD00132725
CAS 11113-61-4
Chemical Name or Material Dowex™ 50WX4 200-400 (H)
Quantity 1000g

Specifications

Chemical Name or Material Dowex™ 50WX4 200-400 (H)
CAS 11113-61-4
MDL Number MFCD00132725

Applications
Dowex® 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex® 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.