Learn More
Dowex™ 50WX4 100-200 (H), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L139170B
Description
Dowex™ 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Dowex™ 50WX4 100-200 (H) | |
MFCD00132725 | |
Insoluble in water. |
11113-61-4 | |
1000 g |
Chemical Identifiers
11113-61-4 | |
MFCD00132725 |
Safety and Handling
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only