missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX2 50-100 (H)

Manufacturer:  Alfa Aesar™ L139430B

 View more versions of this product

Catalog No. AAL139430B


 

Specifications

Description

Dowex 50WX2 50-100 (H); Alfa Aesar; 1000g; CAS: 12612-37-2

Applications
Dowex® 50WX2 50-100 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Description & Specifications

Specifications

Dowex™ 50WX2 50-100 (H)
12612-37-2
MFCD00132722
1000g

Chemical Identifiers

12612-37-2
MFCD00132722
Dowex™ 50WX2 50-100 (H)