Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 50WX2 50-100 (H)


Quantity 250g

Manufacturer: alfa aesar™  L1394330

 View more versions of this product

Catalog No. AAL1394330

  • / Each
 

Description & Specifications

Chemical Identifiers

MDL Number MFCD00132722
CAS 12612-37-2
Chemical Name or Material Dowex™ 50WX2 50-100 (H)
Quantity 250g

Specifications

Chemical Name or Material Dowex™ 50WX2 50-100 (H)
CAS 12612-37-2
MDL Number MFCD00132722

Applications
Dowex® 50WX2 50-100 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.