Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 50-100 (Cl)

$662.47 - $662.47

Chemical Identifiers

Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL142560B

  SDS
alfa aesar™
L142560B
1000g Each for $662.47
Description & Specifications

Specifications

Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
MDL Number MFCD00132718
CAS 12627-85-9
Quantity 1000g
Odor Amine-like

Applications
Dowex|r 1X8 50-100 (Cl) is used in fine chemical processing and as an adsorbent in solid phase separation. It is also used in the determination of pantothenate kinase assay using Dowex column. It is involved in the separation of inositol phosphates from myo-[3H]inositol by Dowex chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.