Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 50-100 (Cl)


Quantity 50g

Manufacturer: alfa aesar™  L1425618

 View more versions of this product

Catalog No. AAL1425618

  • / Each
 

Description & Specifications

Chemical Identifiers

MDL Number MFCD00132718
CAS 12627-85-9
Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
Odor Amine-like
Quantity 50g

Specifications

Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718

Applications
Dowex|r 1X8 50-100 (Cl) is used in fine chemical processing and as an adsorbent in solid phase separation. It is also used in the determination of pantothenate kinase assay using Dowex column. It is involved in the separation of inositol phosphates from myo-[3H]inositol by Dowex chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.