Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 50-100 (Cl)


Quantity 1000g

Manufacturer: alfa aesar™  L14256-0B

 View more versions of this product

Catalog No. AAL142560B

  • / Each
 

Description & Specifications

Chemical Identifiers

Odor Amine-like
Quantity 1000g
Quantity 1000g
Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718

Specifications

Chemical Name or Material Dowex™ 1X8 50-100 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718

Applications
Dowex|r 1X8 50-100 (Cl) is used in fine chemical processing and as an adsorbent in solid phase separation. It is also used in the determination of pantothenate kinase assay using Dowex column. It is involved in the separation of inositol phosphates from myo-[3H]inositol by Dowex chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.