Dowex™ 1X8, 200-400 mesh, ion-exchange resin, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals 203010025
Catalog No. AC203010025
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Dowex™ 1X8
15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh)
C29H34ClN
Cl form: 39 to 45%
01,511; 04,267; 06,302; 07,182; 08,263
2.5kg
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
16212807
12627-85-9
Plastic bottle
MFCD00132715,MFCD00132718
White to Yellow
10, 2182
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
432.05
SDS Documents