Please login to your online account to display your discounted pricing

Dowex™ 1X8-400, ion-exchange resin, ACROS Organics™

$466.05 - $466.05

Chemical Identifiers

Chemical Name or Material Dowex™ 1X8
CAS 12627-85-9
MDL Number MFCD00132718
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203010025

  SDS
thermo fisher scientific
AC203010025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203011000

  SDS
fisher scientific acros
203011000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203015000

  SDS
fisher scientific acros
203015000
Plastic bottle 500g Each for $466.05
Description & Specifications

Specifications

Physical Form Moist Beads
Quantity 2.5kg
Color White to yellow
Merck Index 10, 2182
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Water Volume Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 39 to 45%
Chemical Name or Material Dowex™ 1X8
Particle Size 15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh)
MDL Number MFCD00132718
Packaging Plastic bottle
CAS 12627-85-9