missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 1X8-400, ion-exchange resin, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 203015000

 View more versions of this product

Catalog No. AC203015000


 

Specifications Description & Specifications

Specifications

Dowex™ 1X8
15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh)
Moist Beads
Cl form: 39 to 45%
White to yellow
10, 2182
12627-85-9
Plastic bottle
wet volume capacity, Cl form (meq/mL):> 1.2
MFCD00132718
01,511; 04,267; 06,302; 07,182; 08,263
500g

Chemical Identifiers

12627-85-9
MFCD00132718
Dowex™ 1X8