missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 1X8-400, ion-exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203011000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Moist Beads | |
12627-85-9 | |
Plastic bottle | |
C29H34ClN | |
Cl form: 39 to 45% | |
01,511; 04,267; 06,302; 07,182; 08,263 | |
100 g | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride | |
16212807 |
Dowex™ 1X8 | |
15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh) | |
wet volume capacity, Cl form (meq/mL):> 1.2 | |
MFCD00132718 | |
White to Yellow | |
10, 2182 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
* | |
432.05 |
Chemical Identifiers
12627-85-9 | |
432.05 | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
16212807 |
C29H34ClN | |
MFCD00132718 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
* |
RUO – Research Use Only