missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 1X8-200, ion-exchange resin, ACROS Organics™

Manufacturer:  Acros Organics 203005000

 View more versions of this product

Catalog No. AC203005000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Ion exchange resin
Specifications

Specifications

Dowex™ 1X8
15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
C29H34ClN
wet volume capacity, Cl form (meq/mL):> 1.2
01,511; 04,267; 06,302; 07,182; 08,263
dowex 1x2 50-100 cl
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
16212807
12627-85-9
Plastic bottle
MFCD00132718
Cl form: 39 to 45%
Tan to White
500g
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
432.048
Beads

Chemical Identifiers

12627-85-9
432.048
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
C29H34ClN
MFCD00132718
dowex 1x2 50-100 cl
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
SDS Documents