missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 1X8-200, ion-exchange resin, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 203001000

 View more versions of this product

Catalog No. AC203001000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Beads
White to light yellow to tan
01,511; 04,267; 06,302; 07,182; 08,263
12627-85-9
wet volume capacity, Cl form (meq/mL):> 1.2
Plastic bottle
100g
15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
Dowex™ 1X8
MFCD00132718
Cl form: 39 to 45%

Chemical Identifiers

12627-85-9
MFCD00132718
Dowex™ 1X8