missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dowex™ 1X8-200, ion-exchange resin, ACROS Organics™

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.048
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
SMILES C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC203001000 SDS View Documents Acros Organics
203001000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC203005000 SDS View Documents Acros Organics
203005000
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC203000025 SDS View Documents Acros Organics
AC203000025
2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Ion exchange resin
Specifications

Chemical Identifiers

12627-85-9
432.048
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
C29H34ClN
MFCD00132718
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride

Specifications

12627-85-9
Plastic bottle
MFCD00132718
Cl form: 39 to 45%
Tan to White
100g
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
432.048
Beads
15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
C29H34ClN
wet volume capacity, Cl form (meq/mL):> 1.2
01,511; 04,267; 06,302; 07,182; 08,263
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
16212807
Dowex™ 1X8