missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 1X8, 100-200 mesh, ion-exchange resin, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 203000025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Ion exchange resinSpecifications
Dowex™ 1X8 | |
15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh) | |
MFCD00132715,MFCD00132718 | |
Tan to White | |
2.5kg | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride | |
16212807 |
12627-85-9 | |
C29H34ClN | |
Cl form: 39 to 45% | |
01,511; 04,267; 06,302; 07,182; 08,263 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 | |
432.05 |
RUO â Research Use Only