Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 200-400 (Cl)


Quantity 250g

Manufacturer: alfa aesar™  L1425830

 View more versions of this product

Catalog No. AAL1425830

  • / Each
 

Description & Specifications

Chemical Identifiers

CAS 12627-85-9
Chemical Name or Material Dowex™ 1X8 200-400 (Cl)
Odor Amine-like
MDL Number MFCD00132718
Quantity 250g

Specifications

Chemical Name or Material Dowex™ 1X8 200-400 (Cl)
CAS 12627-85-9
MDL Number MFCD00132718

Applications
Dowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.