missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dowex™ 1X8 200-400 (Cl), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L142580B
Description
Dowex™ 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Dowex™ 1X8 200-400 (Cl) | |
Amine-like | |
MFCD00132718 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride | |
16212807 |
12627-85-9 | |
C29H34ClN | |
1000 g | |
Insoluble in water. | |
* | |
432.05 |
Chemical Identifiers
12627-85-9 | |
432.05 | |
BBQMUEOYPPPODD-UHFFFAOYSA-M | |
16212807 |
C29H34ClN | |
MFCD00132718 | |
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
* |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only