missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dowex™ 1X8 200-400 (Cl)

Manufacturer:  Alfa Aesar™ L1425818

 View more versions of this product

Catalog No. AAL1425818



Description

Description

Applications
Dowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Dowex™ 1X8 200-400 (Cl)
Amine-like
MFCD00132718
50g
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
432.048
12627-85-9
C29H34ClN
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
16212807

Chemical Identifiers

12627-85-9
432.048
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
C[N+](C)(C)C1=CC=C(C=C1)C=C.C=CC1=CC=CC=C1.C=CC1=CC=C(C=C1)C=C.[Cl-]
C29H34ClN
MFCD00132718
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
Documents